Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Acetyl-2-hydroxybenzamide, 97%, Thermo Scientific™

Catalog No. SB01472FA
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB01472FA 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SB01472FA Supplier Thermo Scientific Chemicals Supplier No. SB01472FA

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 40187-51-7
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.175
InChI Key LWAQTCWTCCNHJR-UHFFFAOYSA-N
Synonym 5-acetylsalicylamide, benzamide, 5-acetyl-2-hydroxy, 5-acetyl-2-hydroxy-benzamide, 5-acetyl salicylamide, acmc-1alb7, dsstox_cid_31525, dsstox_rid_97410, dsstox_gsid_57736, 4-10-00-03635 beilstein handbook reference
PubChem CID 198212
IUPAC Name 5-acetyl-2-hydroxybenzamide
SMILES CC(=O)C1=CC(=C(C=C1)O)C(=O)N

Specifications

Assay Percent Range 97%
Quantity 100 g
Formula Weight 179.17
Chemical Name or Material 5-Acetyl-2-hydroxybenzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.