Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-Acetyl-8-(benzyloxy)-1,2-dihydroquinolin-2-one

Catalog No. KOB00281G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
KOB00281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. KOB00281G Supplier Sigma Aldrich Supplier No. KOB00281G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Methyl 4-iodobutyrate can be synthesized from methyl-4-chlorobutyrate and sodium iodide.methyl 4-iodobutyrate may be used as reagent in the synthesis of 4-[formyl-5-(methoxymethyl)-1h-pyrrol-1-yl] butanoic acid
  • It has been used in the synthesis of a range of stable s-adenosylmethionine (sam) mimetics
  • Their ability to promote the binding of the e
  • Coli methionine repressor (metj) to its operator DNA has been investigated

Specifications

CAS 93609-84-8
Molecular Formula C18H15NO3
Linear Formula C18H15NO3
MDL Number MFCD19381743
Quantity 1 g
Molecular Weight (g/mol) 293.32
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.