Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Acetylsalicylamide 98.0+%, TCI America™
SDP

Catalog No. A115625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A115625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A115625G Supplier TCI America Supplier No. A115625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 40187-51-7
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.175
MDL Number MFCD00049222
InChI Key LWAQTCWTCCNHJR-UHFFFAOYSA-N
Synonym 5-acetylsalicylamide, benzamide, 5-acetyl-2-hydroxy, 5-acetyl-2-hydroxy-benzamide, 5-acetyl salicylamide, acmc-1alb7, dsstox_cid_31525, dsstox_rid_97410, dsstox_gsid_57736, 4-10-00-03635 beilstein handbook reference
PubChem CID 198212
IUPAC Name 5-acetyl-2-hydroxybenzamide
SMILES CC(=O)C1=CC(=C(C=C1)O)C(=O)N

Specifications

Melting Point 224°C
Color White-Yellow
Quantity 25 g
Formula Weight 179.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Acetylsalicylamide
RTECSNumber CU8702280
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.