Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Acetylthiophene-2-carbonitrile, 97%, Thermo Scientific™

Catalog No. TL00354EA
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
TL00354EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TL00354EA Supplier Thermo Scientific Chemicals Supplier No. TL00354EA

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 88653-55-8
Molecular Formula C7H5NOS
Molecular Weight (g/mol) 151.183
InChI Key VSHPLUBHIUFLES-UHFFFAOYSA-N
Synonym 2-acetyl-5-cyanothiophene, 2-acetyl-5-cyanotiophene, 5-cyano-2-acetylthiophene, 5-acetyl-2-thiophenecarbonitrile, maybridge1_000101, acmc-20ao65, 2-cyano-5-acetyl-thiophene, 5-acetyl-thiophene-2-carbonitrile, 5-ethanoylthiophene-2-carbonitrile, 2-thiophenecarbonitrile, 5-acetyl-7ci,9ci
PubChem CID 2747565
IUPAC Name 5-acetylthiophene-2-carbonitrile
SMILES CC(=O)C1=CC=C(S1)C#N

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 151.19
Percent Purity 97%
Chemical Name or Material 5-Acetylthiophene-2-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.