Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Acetylthiophene-2-carbonitrile 98.0+%, TCI America™
SDP

Catalog No. A274625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A274625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A274625G Supplier TCI America Supplier No. A274625G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 88653-55-8
Molecular Formula C7H5NOS
Molecular Weight (g/mol) 151.183
MDL Number MFCD00052314
InChI Key VSHPLUBHIUFLES-UHFFFAOYSA-N
Synonym 2-acetyl-5-cyanothiophene, 2-acetyl-5-cyanotiophene, 5-cyano-2-acetylthiophene, 5-acetyl-2-thiophenecarbonitrile, maybridge1_000101, acmc-20ao65, 2-cyano-5-acetyl-thiophene, 5-acetyl-thiophene-2-carbonitrile, 5-ethanoylthiophene-2-carbonitrile, 2-thiophenecarbonitrile, 5-acetyl-7ci,9ci
PubChem CID 2747565
IUPAC Name 5-acetylthiophene-2-carbonitrile
SMILES CC(=O)C1=CC=C(S1)C#N

Specifications

Melting Point 79°C
Color Yellow
Boiling Point 97°C
Quantity 25 g
UN Number 3439
Formula Weight 151.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Acetylthiophene-2-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.