Learn More
Cayman Chemical 5-AEDBhydrochlorIde 10mg
SDP
Catalog No. 502967888
Encompass_Preferred
An analytical reference standard categorized as a benzofuran; has been found in samples seized by law enforcement; intended for research and forensic applications
Catalog No. 50-296-7888 Supplier Cayman Chemical Supplier No. 1835110 MG
An analytical reference standard categorized as a benzofuran; has been found in samples seized by law enforcement; intended for research and forensic applications