Learn More
Enzo Life Sciences 5-AIQ. hydrochloride (5mg). CAS: 93117-07-8
SDP
Supplier:  Enzo Life Sciences ALX270285M005
Encompass_Preferred
Water-soluble, potent inhibitor of poly(ADP-ribose) polymerase-1 (PARP-1). Alternative name: 5-Aminoisoquinolinone . HCl. Formula: C9H8N2O . HCl. MW: 160.2 . 36.5. Purity: ≥97% (HPLC). Solubility: Soluble in water (warm). Long Term Storage: +4°C. Handling: Protect from moisture.
Catalog No. 50-200-8463