Learn More
Medchemexpress LLC HY-133100 10mg Medchemexpress, 5-ALA benzyl ester (hydrochloride) CAS:163271-32-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13310010MG
Encompass_Preferred
Medchemexpress, HY-133100 10mg 5-ALA benzyl ester (HCl) CAS:163271-32-7 5-ALA benzyl ester hydrochloride (Benzyl-ALA hydrochloride) is a protoporphyrin precursor used as a photodetection agent. 5-ALA benzyl ester hydrochloride induces protoporphyrin IX (PPIX) accumulation in colon carcinoma cell lines. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9850