Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Allyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamate 96.0+%, TCI America™
SDP

Catalog No. A28955G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28955G 5 g
A28951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28955G Supplier TCI America Supplier No. A28955G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 133464-46-7
Molecular Formula C23H23NO6
Molecular Weight (g/mol) 409.44
MDL Number MFCD00190879
InChI Key LRBARFFNYOKIAX-UHFFFAOYNA-N
Synonym fmoc-glu oall-oh, fmoc-l-glutamic acid 5-allyl ester, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-allyloxy-5-oxopentanoic acid, ambotzfaa1372, 5-allyl n-fmoc-l-glutamate, n-fmoc-l-glutamic acid 5-allyl ester, fmoc-glu oall-oh hplc, 5-allyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate, n-9h-fluorene-9-ylmethoxycarbonyl-l-glutamic acid 5-allyl ester, n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamic acid 5-allyl ester
PubChem CID 7019067
IUPAC Name 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxo-5-(prop-2-en-1-yloxy)pentanoic acid
SMILES OC(=O)C(CCC(=O)OCC=C)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 127°C
Color White
Quantity 5 g
Formula Weight 409.44
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Allyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.