Learn More
Medchemexpress LLC 5-amino-1,3,4-thiadiazole-2-sulfonamide | 14949-00-9 | 98.0% | 180.21 g·mol⁻¹ | C2H4N4O2S2 | 5 MG
SDP
Catalog No. 5000210457
Encompass_Preferred
CL 5343 (5-amino-1,3,4-thiadiazole-2-sulfonamide) is a small-molecule inhibitor of carbonic anhydrases, selectively inhibiting human carbonic anhydrase B (HCA-B) and isoforms I, II, IV, and VII. It is supplied for research use to support biochemical assays and early drug discovery studies involving carbonic anhydrase pathways.
  • selective carbonic anhydrase inhibition profile
  • documented molecular formula and molecular weight for assay planning
  • available in small research pack sizes for assay development
  • soluble in common solvents for stock solution preparation
  • intended for research use only; not for therapeutic use
Catalog No. 50-002-10457 Supplier Medchemexpress LLC Supplier No. HYW0749755MG
CL 5343 (5-amino-1,3,4-thiadiazole-2-sulfonamide) is a small-molecule inhibitor of carbonic anhydrases, selectively inhibiting human carbonic anhydrase B (HCA-B) and isoforms I, II, IV, and VII. It is supplied for research use to support biochemical assays and early drug discovery studies involving carbonic anhydrase pathways.
  • selective carbonic anhydrase inhibition profile
  • documented molecular formula and molecular weight for assay planning
  • available in small research pack sizes for assay development
  • soluble in common solvents for stock solution preparation
  • intended for research use only; not for therapeutic use