Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-amino-1,3,4-thiadiazole-2-sulfonamide | 14949-00-9 | MFCD03425417 | 98.0% | 180.21 g/mol | C2H4N4O2S2 | 100 MG
SDP

Catalog No. 5000209797
Encompass_Preferred

CL 5343 (5-amino-1,3,4-thiadiazole-2-sulfonamide) is a small-molecule carbonic anhydrase inhibitor supplied for research use. It is used in biochemical assays targeting carbonic anhydrase isoforms and as a ligand in targeted delivery studies.

  • Acts as a carbonic anhydrase inhibitor.
  • CAS number 14949-00-9.
  • Molecular formula C2H4N4O2S2.
  • Molecular weight 180.21 g/mol.
  • Purity 98.0%.
  • Available as a solid in common laboratory pack sizes, including 100 MG.

Catalog No. 50-002-09797 Supplier Medchemexpress LLC Supplier No. HYW074975100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CL 5343 (5-amino-1,3,4-thiadiazole-2-sulfonamide) is a small-molecule carbonic anhydrase inhibitor supplied for research use. It is used in biochemical assays targeting carbonic anhydrase isoforms and as a ligand in targeted delivery studies.

  • Acts as a carbonic anhydrase inhibitor.
  • CAS number 14949-00-9.
  • Molecular formula C2H4N4O2S2.
  • Molecular weight 180.21 g/mol.
  • Purity 98.0%.
  • Available as a solid in common laboratory pack sizes, including 100 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.