Learn More
Medchemexpress LLC 5-amino-1,3,4-thiadiazole-2-sulfonamide | 14949-00-9 | MFCD03425417 | 98.0% | 180.21 g/mol | C2H4N4O2S2 | 25 MG
SDP
Catalog No. 5000209084
Encompass_Preferred
A small-molecule carbonic anhydrase inhibitor supplied as a white to off-white solid for in vitro research. The compound is soluble in DMSO and is provided in multiple sizes for biochemical and cell-based assays.
  • solid form suitable for analytical and biological assays
  • available as a 10 mM solution in DMSO for direct use in assays
  • high purity (98.0%) for reliable activity data
  • compact packaging options including 25 mg for small-scale experiments
  • stable under recommended storage conditions when protected from light
Catalog No. 50-002-09084 Supplier Medchemexpress LLC Supplier No. 25MGHYW074975
A small-molecule carbonic anhydrase inhibitor supplied as a white to off-white solid for in vitro research. The compound is soluble in DMSO and is provided in multiple sizes for biochemical and cell-based assays.
  • solid form suitable for analytical and biological assays
  • available as a 10 mM solution in DMSO for direct use in assays
  • high purity (98.0%) for reliable activity data
  • compact packaging options including 25 mg for small-scale experiments
  • stable under recommended storage conditions when protected from light