Learn More
Accela Chembio Inc 5-amino-1-ethyl-1h-indazole | 1g | 511249-17-5 | MFCD16620004 | 95+% | Shelf Life: 1260 Days | Light Sensitive/nitrogen Or Argon/+4
Supplier:  Accela Chembio Inc SY0066671G
Encompass_Preferred
5-amino-1-ethyl-1h-indazole | 1g | 511249-17-5 | MFCD16620004 | 95+% | Shelf Life: 1260 Days | Light Sensitive/nitrogen Or Argon/+4
Catalog No. 50-354-535