Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5-Amino-1-methyl-1H-indazole, 97%, Thermo Scientific™
Specifications
Specifications
| CAS Min % | 96.0 |
| CAS Max % | 100.0 |
| Melting Point | 152.0°C to 154.0°C |
| Color | Brown |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97% min. (HPLC) |
| Packaging | Glass bottle |
| Quantity | 1 g |
| Formula Weight | 147.18 |
| Percent Purity | 97% |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes serious eye irritation. Harmful if swallowed. May cause respiratory irritation. Harmful in contact with skin. Causes skin irritation. Harmful if inhaled. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF |
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?