Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-1-methyl-1H-indazole, 97%, Thermo Scientific™

Catalog No. AC429520050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC429520050 5 g Glass bottle
AC429520010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC429520050 Supplier Thermo Scientific Chemicals Supplier No. 429520050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 50593-24-3
Molecular Formula C8H9N3
Molecular Weight (g/mol) 147.18
MDL Number MFCD03305455
InChI Key PYOFNPHTKBSXOM-UHFFFAOYSA-N
Synonym 1-methyl-1h-indazol-5-amine, 5-amino-1-methyl-1h-indazole, 1-methyl-1h-indazol-5-ylamine, 1h-indazol-5-amine, 1-methyl, 5-amino-1-methylindazole, 1-methyl-1h-indazole-5-amine, pubchem20587, 1-methyl-5-aminoindazol, 1-methyl-5-indazolamine, 1-methyl-5-aminoindazole
PubChem CID 2768032
IUPAC Name 1-methylindazol-5-amine
SMILES CN1C2=C(C=C(C=C2)N)C=N1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 152.0°C to 154.0°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 97% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 147.18
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-1-methyl-1H-indazole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.