Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-Amino-1-pentanol

Catalog No. 12304810G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-304-810G 10 g
12-304-850G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-304-810G Supplier Sigma Aldrich Supplier No. 12304810G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

5-Amino-1-pentanol

Specifications

CAS 2508-29-4
Melting Point 33°C to 35°C (lit.)
Density 0.949 g/mL (at 25°C (literature))
Boiling Point 120°C to 122°C (16 mmHg)
Molecular Formula C5H13NO
Refractive Index n20/D 1.4615 (literature)
Linear Formula NH2(CH2)5 OH
MDL Number MFCD00008237
Quantity 10 g
Molecular Weight (g/mol) 103.16
Percent Purity ≥92.0% (GC)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.