Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2,2,4-trimethyl-1-cyclopentanemethylamine, Mixture of Isomers 99%, Thermo Scientific™

Catalog No. AC186922500
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC186922500 250 g
AC186920010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC186922500 Supplier Thermo Scientific Chemicals Supplier No. 186922500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67907-32-8
Molecular Formula C9H20N2
Molecular Weight (g/mol) 156.273
InChI Key QKHWUKPTSMULMZ-UHFFFAOYSA-N
Synonym 2-aminomethyl-3,3,5-trimethylcyclopentanamine, cyclopentanemethanamine, 5-amino-2,2,4-trimethyl, 1-amino-2-aminomethyl-3,3,5-trimethylcyclopentane, 5-amino-2,2,4-trimethyl-1-cyclopentanemethylamine, 2-aminomethyl-3,3,5-trimethylcyclopentan-1-amine, 5-aminomethyl-2,4,4-trimethyl-1-cyclopentylamine, 5-amino-2,2,4-trimethylcyclopentylmethylamine, 5-amino-2,2,4-trimethylcyclopentanemethanamine, 2-aminomethyl-3,3,5-trimethylcyclopentanamine #, 5-amino-2,2,4-trimethyl-1-cyclopentanemethylamine, mixture of isomers
PubChem CID 106962
IUPAC Name 2-(aminomethyl)-3,3,5-trimethylcyclopentan-1-amine
SMILES CC1CC(C(C1N)CN)(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Refractive Index 1.4730 to 1.4750 (20°C, 589nm)
Linear Formula H2NC5H5(CH3)3CH2NH2
Quantity 250 g
Formula Weight 156.27
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material 5-Amino-2,2,4-trimethyl-1-cyclopentanemethylamine, mixture of isomers, 99%
Show More Show Less
EINECSNumber 267-741-1
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.