Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-amino-2-(2,6-diisopropylphenyl)isoindoline-1,3-dione | 100823-03-8 | MFCD03085319 | 99.6% | 322.40 g/mol | C20H22N2O2 | 5 MG
SDP

Catalog No. 5000262083
Encompass_Preferred

TC11 is a research-grade small molecule that functions as an MCL1 degrader and activator of caspase-9 and CDK1. It is a phenylacetylamide derivative that promotes MCL1 degradation and induces apoptotic death during prolonged mitotic arrest; supplied for laboratory research use only.

  • High purity (99.6%).
  • Molecular weight 322.40 g/mol.
  • Chemical formula C20H22N2O2.
  • Solid, white to yellow appearance.
  • Store at 4°C and protect from light; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Functions as an MCL1 degrader and caspase-9 and CDK1 activator in cell studies.
  • Intended for research use only; not for human or clinical applications.

Catalog No. 50-002-62083 Supplier Medchemexpress LLC Supplier No. HY1294785MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TC11 is a research-grade small molecule that functions as an MCL1 degrader and activator of caspase-9 and CDK1. It is a phenylacetylamide derivative that promotes MCL1 degradation and induces apoptotic death during prolonged mitotic arrest; supplied for laboratory research use only.

  • High purity (99.6%).
  • Molecular weight 322.40 g/mol.
  • Chemical formula C20H22N2O2.
  • Solid, white to yellow appearance.
  • Store at 4°C and protect from light; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Functions as an MCL1 degrader and caspase-9 and CDK1 activator in cell studies.
  • Intended for research use only; not for human or clinical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.