Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2,4,6-triiodoisophthalic acid, 95%

Catalog No. AAB2217818
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 35453-19-1 | C8H4I3NO4 | 558.836 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35453-19-1
Molecular Formula C8H4I3NO4
Molecular Weight (g/mol) 558.836
MDL Number MFCD00190167
InChI Key JEZJSNULLBSYHV-UHFFFAOYSA-N
Synonym 5-amino-2,4,6-triiodoisophthalic acid, unii-a17g990rt0, 5-amino-2,4,6-triiodo-1,3-benzenedicarboxylic acid, 5-amino-2,4,6-iodoisophthalic acid, 1,3-benzenedicarboxylic acid, 5-amino-2,4,6-triiodo, 5-amino-2,4,6-triiodoisophthalicacid, isophthalic acid, 5-amino-2,4,6-triiodo, acmc-209ih0, ksc224s6b, aronis25361
PubChem CID 3015783
IUPAC Name 5-amino-2,4,6-triiodobenzene-1,3-dicarboxylic acid
SMILES C1(=C(C(=C(C(=C1I)N)I)C(=O)O)I)C(=O)O

Specifications

Melting Point 265°C to 270°C
Quantity 50 g
Sensitivity Light and moisture sensitive
Formula Weight 558.84
Percent Purity 95%
Chemical Name or Material 5-Amino-2,4,6-triiodoisophthalic acid
Hazard Category H317-H334
Hazard Statement GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
EINECSNumber 252-575-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.