Learn More
Medchemexpress LLC 5-Amino-2-fluoropyridine | 1827-27-6 | 99.9% | C5H5FN2 | 10 G
SDP
Catalog No. 5000433790
Encompass_Preferred
5-Amino-2-fluoropyridine is a fluorinated heterocyclic compound used for the synthesis of other active compounds. Its amino and fluorine atoms contribute to its important value in biological activity research, such as enzyme inhibition and antibacterial effects, and in the preparation of functional materials.
  • Fluorinated heterocyclic compound
  • Used for synthesis of active compounds
  • Valuable in enzyme inhibition research
  • Valuable in antibacterial effects research
  • Valuable in preparation of functional materials
Catalog No. 50-004-33790 Supplier Medchemexpress LLC Supplier No. HY6525410G
5-Amino-2-fluoropyridine is a fluorinated heterocyclic compound used for the synthesis of other active compounds. Its amino and fluorine atoms contribute to its important value in biological activity research, such as enzyme inhibition and antibacterial effects, and in the preparation of functional materials.
  • Fluorinated heterocyclic compound
  • Used for synthesis of active compounds
  • Valuable in enzyme inhibition research
  • Valuable in antibacterial effects research
  • Valuable in preparation of functional materials