Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2-(hydroxymethyl)benzeneboronic acid hemiester hydrochloride, 95%

Catalog No. AAH5262903
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5262903 1 g
AAH52629MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5262903 Supplier Thermo Scientific Chemicals Supplier No. H5262903
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 117098-93-8 | C7H9BClNO2 | 185.41 g/mol

5-Amino-2-(hydroxymethyl)benzeneboronic acid hemiester hydrochloride is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Amino-2-(hydroxymethyl)benzeneboronic acid hemiester hydrochloride is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117098-93-8
Molecular Formula C7H9BClNO2
Molecular Weight (g/mol) 185.41
MDL Number MFCD04115645
InChI Key ZDCBDYGPSUVCOU-UHFFFAOYSA-N
Synonym 6-aminobenzo c 1,2 oxaborol-1 3h-ol hydrochloride, 6-amino-1-hydroxy-2,1-benzoxaborolane hydrochloride, 5-amino-2-hydroxymethylphenylboronic acid, hcl, dehydrate, 5-amino-2-hydroxymethylphenyl boronic acid, hcl, dehydrate, 6-amino-3h-2,1-benzoxaborol-1-ol hydrochloride, 1-hydroxy-3h-2,1-benzoxaborol-6-amine hydrochloride, 5-amino-2-hydroxymethylphenyl boronic acid, hydrochloride, dehydrate, 6-amino-1-hydroxy-2,1-benzoxaborolane, hcl, 6-amino-2,1-benzoxaborol-1 3h-ol-hydrogen chloride 1/1, 5-amino-2-hydroxymethylphenyl boronicacid, hcl, dehydrate
PubChem CID 44118730
IUPAC Name 1-hydroxy-3H-2,1-benzoxaborol-6-amine;hydrochloride
SMILES Cl.NC1=CC2=C(COB2O)C=C1

Specifications

Melting Point 300°C
Quantity 1 g
Formula Weight 185.42
Percent Purity 95%
Chemical Name or Material 5-Amino-2-(hydroxymethyl)benzeneboronic acid hemiester hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.