Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Amino-2-methoxypyridine | 6628-77-9 | 124.14 | 25 G
SDP

Catalog No. 5000448185
Encompass_Preferred

5-Amino-2-methoxypyridine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is stable under recommended storage conditions and is suitable for various laboratory applications.

  • Used as a biochemical reagent
  • Target biochemical assay reagents
  • Soluble in DMSO at 100 mg/mL
  • Store solid at 4°C, protect from light
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protect from light

Catalog No. 50-004-48185 Supplier Medchemexpress LLC Supplier No. 25GHYW001994
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Amino-2-methoxypyridine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is stable under recommended storage conditions and is suitable for various laboratory applications.

  • Used as a biochemical reagent
  • Target biochemical assay reagents
  • Soluble in DMSO at 100 mg/mL
  • Store solid at 4°C, protect from light
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protect from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.