Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5'-Amino-2'-methylacetanilide 98.0+%, TCI America™
SDP

Catalog No. A27241G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27241G 1 g
A27245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27241G Supplier TCI America Supplier No. A27241G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5434-30-0
Molecular Formula C9H12N2O
Molecular Weight (g/mol) 164.208
MDL Number MFCD00047859
InChI Key UAZGSMMESOKKQZ-UHFFFAOYSA-N
Synonym n-5-amino-2-methylphenyl acetamide, 2-acetylamino-4-aminotoluene, acetamide, n-5-amino-2-methylphenyl, 5'-amino-2'-methylacetanilide, 2-acetylamino-p-toluidine, n1-5-amino-2-methylphenyl acetamide, n-3-amino-6-methylphenyl acetamide, 3-acetylamino-p-toluidine, 2-methyl-5-aminoacetaniline, 3-acetamido-4-methylaniline
PubChem CID 79492
IUPAC Name N-(5-amino-2-methylphenyl)acetamide
SMILES CC1=C(C=C(C=C1)N)NC(=O)C

Specifications

Melting Point 144°C
Color White-Yellow
Quantity 1 g
Formula Weight 164.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5′-Amino-2′-methylacetanilide
EINECSNumber (3)-0270
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.