Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-Amino-2-phenethylisoindoline-1,3-dione

Catalog No. OTV0008081G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV0008081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV0008081G Supplier Sigma Aldrich Supplier No. OTV0008081G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2,6,6-trimethyl-1-cyclohexene-1-acetaldehyde can be used as a starting material to prepare:
  • (+/-)-aeginetolide by oxidation in the presence of meta-chloroperoxybenzoic acid (m-cpba)
  • (+/-)-dihydroactinidiolide (a c11-terpenic lactone) via dehydration of key intermediate aeginetolide. it can also be used as a key intermediate to synthesize drimane-related sesquiterpenes and substituted retinoic acid analogs.contains beta-cyclocitral

Specifications

Molecular Formula C16H14N2O2
Linear Formula C16H14N2O2
MDL Number MFCD12448364
Quantity 1 g
Molecular Weight (g/mol) 266.29
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.