Learn More
Description
- 1,1,1-trifluoro-3-phenyl-2-propanone is a trifluoromethyl ketone
- Its in vitro neuroprotective potency against low k(+)-induced apoptosis in cerebellar granule neurons (cgns) by different pathways has been reported
Specifications
Specifications
| Molecular Formula | C5H6N2O |
| Linear Formula | C5H6N2O |
| MDL Number | MFCD03427652 |
| Quantity | 5 g |
| Molecular Weight (g/mol) | 110.11 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.