Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Amino-2-((tert-butoxycarbonyl)amino)-5-oxopentanoic acid | 61348-28-5 | 98.0% | 246.26 | 500 G
SDP

Catalog No. 5000452739
Encompass_Preferred

5-Amino-2-((tert-butoxycarbonyl)amino)-5-oxopentanoic acid is a glutamine derivative. Amino acids and amino acid derivatives are commercially used as ergogenic supplements, influencing anabolic hormone secretion, fuel supply during exercise, mental performance during stress, and preventing exercise-induced muscle damage. They are recognized as beneficial ergogenic dietary substances.

  • Formula: C10H18N2O5
  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Solubility (in vitro): DMSO: 200 mg/mL (812.15 mM; requires ultrasonic; hygroscopic DMSO has a significant impact on product solubility, use newly opened DMSO)

Catalog No. 50-004-52739 Supplier Medchemexpress LLC Supplier No. HYW011983500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Amino-2-((tert-butoxycarbonyl)amino)-5-oxopentanoic acid is a glutamine derivative. Amino acids and amino acid derivatives are commercially used as ergogenic supplements, influencing anabolic hormone secretion, fuel supply during exercise, mental performance during stress, and preventing exercise-induced muscle damage. They are recognized as beneficial ergogenic dietary substances.

  • Formula: C10H18N2O5
  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Solubility (in vitro): DMSO: 200 mg/mL (812.15 mM; requires ultrasonic; hygroscopic DMSO has a significant impact on product solubility, use newly opened DMSO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.