Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-4-bromo-3-methyl-1-phenylpyrazole 96.0+%, TCI America™
SDP

Catalog No. A29015G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29015G 5 g
A29011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29015G Supplier TCI America Supplier No. A29015G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 69464-98-8
Molecular Formula C10H10BrN3
Molecular Weight (g/mol) 252.115
MDL Number MFCD04039926
InChI Key KNUGDEKPCNCMIO-UHFFFAOYSA-N
Synonym 4-bromo-3-methyl-1-phenyl-1h-pyrazol-5-amine, 4-bromo-3-methyl-1-phenyl-1h-pyrazol-5-ylamine, 5-amino-4-bromo-3-methyl-1-phenylpyrazole, acmc-20amn6, 4-bromo-5-methyl-2-phenyl-2h-pyrazol-3, 5-amino-4-bromo-3-methyl-1-phenyl-1h-pyrazole, 4-bromo-5-methyl-2-phenyl-2h-pyrazol-3-ylamine, 4-bromo-3-methyl-1-phenyl-1h-pyrazol-5-, 1h-pyrazol-5-amine, 4-bromo-3-methyl-1-phenyl
PubChem CID 4339679
IUPAC Name 4-bromo-5-methyl-2-phenylpyrazol-3-amine
SMILES CC1=NN(C(=C1Br)N)C2=CC=CC=C2

Specifications

Melting Point 104°C
Color Yellow
Quantity 5 g
Formula Weight 252.12
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-4-bromo-3-methyl-1-phenylpyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.