Learn More
TARGETMOL CHEMICALS INC 5-Amino-5-deoxyadenosine 5MG
SDP
Catalog No. 502877452
Encompass_Preferred
Also available in 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. 5'-Amino-5'-deoxyadenosine (NH2dAdo) is an adenosine kinase inhibitor targeting malignant tumors of the inert lymphatic system with antitumor and anticancer effects. Its mechanism is mediated by the inhibition of DNA synthesis and induction of apoptosis. Purity 95.55%
Catalog No. 50-287-7452 Supplier TARGETMOL CHEMICALS INC Supplier No. T210175MG
Also available in 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. 5'-Amino-5'-deoxyadenosine (NH2dAdo) is an adenosine kinase inhibitor targeting malignant tumors of the inert lymphatic system with antitumor and anticancer effects. Its mechanism is mediated by the inhibition of DNA synthesis and induction of apoptosis. Purity 95.55%