Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-amino-6-chloro-n-[(1-isobutylpiperidin-4-yl)methyl]-2-methylimidazo[1,2-a]pyridine-8-carboxamide | 519148-48-2 | 98.3% | 377.91 g/mol | C19H28ClN5O | 200 MG
SDP

Catalog No. 5000246635
Encompass_Preferred

CJ033466 is a research-grade selective 5-HT4 receptor partial agonist with a reported EC50 of 9 nM and demonstrated gastroprokinetic activity. It is supplied as a purified solid intended for in vitro and in vivo pharmacology research.

  • Selective 5-HT4 receptor partial agonist (EC50 9 nM).
  • Reported gastroprokinetic activity in preclinical studies.
  • High purity (98.29%), suitable for pharmacology research.
  • Molecular formula C19H28ClN5O; molecular weight 377.91 g/mol.
  • Soluble in DMSO (50 mg/mL) with ultrasonic/warming up to 80°C.
  • Storage: solid at 4°C; in solvent -80°C (6 months), -20°C (1 month).
  • Available in research quantities including 200 mg.

Catalog No. 50-002-46635 Supplier Medchemexpress LLC Supplier No. HY103108200MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CJ033466 is a research-grade selective 5-HT4 receptor partial agonist with a reported EC50 of 9 nM and demonstrated gastroprokinetic activity. It is supplied as a purified solid intended for in vitro and in vivo pharmacology research.

  • Selective 5-HT4 receptor partial agonist (EC50 9 nM).
  • Reported gastroprokinetic activity in preclinical studies.
  • High purity (98.29%), suitable for pharmacology research.
  • Molecular formula C19H28ClN5O; molecular weight 377.91 g/mol.
  • Soluble in DMSO (50 mg/mL) with ultrasonic/warming up to 80°C.
  • Storage: solid at 4°C; in solvent -80°C (6 months), -20°C (1 month).
  • Available in research quantities including 200 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.