Learn More
Medchemexpress LLC 5-Amino-8-hydroxyquinoline | 13207-66-4 | 160.18 | 10 MM * 1 ML
SDP
Catalog No. 5000449764
Encompass_Preferred
5-Amino-8-hydroxyquinoline (5A8HQ; 5AHQ) is an orally active non-competitive 20S proteasome inhibitor. It can inhibit NF-κB activity and induce cancer cell death, showing low cytotoxicity towards normal hematopoietic cells. This compound is used for the research of various cancers, such as leukemia.
  • Orally active non-competitive 20S proteasome inhibitor
  • Inhibits NF-κB activity
  • Induces cancer cell death
  • Low cytotoxicity towards normal hematopoietic cells
  • Suitable for cancer research, including leukemia
Catalog No. 50-004-49764 Supplier Medchemexpress LLC Supplier No. HYW00102310MM/1ML
5-Amino-8-hydroxyquinoline (5A8HQ; 5AHQ) is an orally active non-competitive 20S proteasome inhibitor. It can inhibit NF-κB activity and induce cancer cell death, showing low cytotoxicity towards normal hematopoietic cells. This compound is used for the research of various cancers, such as leukemia.
  • Orally active non-competitive 20S proteasome inhibitor
  • Inhibits NF-κB activity
  • Induces cancer cell death
  • Low cytotoxicity towards normal hematopoietic cells
  • Suitable for cancer research, including leukemia