Learn More
Medchemexpress LLC 5-Amino-8-hydroxyquinoline | 13207-66-4 | 160.18 | 50 MG
SDP
Catalog No. 5000447648
Encompass_Preferred
5-Amino-8-hydroxyquinoline (5A8HQ; 5AHQ) is an orally active non-competitive 20S proteasome inhibitor. It can inhibit NF-κB activity and induce cancer cell death, while exhibiting low cytotoxicity towards normal hematopoietic cells. This compound is utilized for cancer research, particularly in studies related to leukemia.
  • Orally active non-competitive 20S proteasome inhibitor
  • Inhibits NF-κB activity
  • Induces cancer cell death
  • Low cytotoxicity towards normal hematopoietic cells
  • Suitable for cancer research, including leukemia studies
Catalog No. 50-004-47648 Supplier Medchemexpress LLC Supplier No. HYW00102350MG
5-Amino-8-hydroxyquinoline (5A8HQ; 5AHQ) is an orally active non-competitive 20S proteasome inhibitor. It can inhibit NF-κB activity and induce cancer cell death, while exhibiting low cytotoxicity towards normal hematopoietic cells. This compound is utilized for cancer research, particularly in studies related to leukemia.
  • Orally active non-competitive 20S proteasome inhibitor
  • Inhibits NF-κB activity
  • Induces cancer cell death
  • Low cytotoxicity towards normal hematopoietic cells
  • Suitable for cancer research, including leukemia studies