Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-amino-n-(2,6-difluorophenyl)-3-(4-sulfamoylanilino)-1,2,4-triazole-1-carbothioamide | 443798-47-8 | 425.44 g·mol⁻1 | C15H13F2N7O2S2 | 5 MG
SDP

Catalog No. 5000245193
Encompass_Preferred

This small-molecule research inhibitor targets cyclin-dependent kinases CDK1 and CDK2, with reported activity against CDC2-like kinases (CLK1 and CLK3). It is provided for biochemical and cellular research into cell-cycle regulation and kinase signaling. Handle with appropriate safety precautions; not for human or veterinary use.

  • Potent CDK1 and CDK2 inhibitor with low-nanomolar activity.
  • Also inhibits CDC2-like kinases (CLK1 and CLK3).
  • Molecular formula C15H13F2N7O2S2, molecular weight 425.44 g·mol⁻1.
  • Supplied as a solid for biochemical and cellular research applications.
  • Storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent store at -80°C (up to 2 years).
  • For research use only; not for human or veterinary use.

Catalog No. 50-002-45193 Supplier Medchemexpress LLC Supplier No. HY1036475MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This small-molecule research inhibitor targets cyclin-dependent kinases CDK1 and CDK2, with reported activity against CDC2-like kinases (CLK1 and CLK3). It is provided for biochemical and cellular research into cell-cycle regulation and kinase signaling. Handle with appropriate safety precautions; not for human or veterinary use.

  • Potent CDK1 and CDK2 inhibitor with low-nanomolar activity.
  • Also inhibits CDC2-like kinases (CLK1 and CLK3).
  • Molecular formula C15H13F2N7O2S2, molecular weight 425.44 g·mol⁻1.
  • Supplied as a solid for biochemical and cellular research applications.
  • Storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent store at -80°C (up to 2 years).
  • For research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.