Learn More
Medchemexpress LLC 5-amino-N-(3-fluoro-4-methoxybenzyl)-3,4-dimethylthieno[2,3-c]pyridazine-6-carboxamide | 1451994-10-7 | 99.3% | 50 MG
SDP
Catalog No. 5000234776
Encompass_Preferred
VU0467485 (AZ13713945) is a potent, selective, orally bioavailable positive allosteric modulator of the muscarinic acetylcholine receptor M4, provided as a research reagent for receptor pharmacology and preclinical studies.
  • Potent and selective M4 positive allosteric modulator.
  • Orally bioavailable properties support in vivo evaluation.
  • High purity suitable for pharmacological assays.
  • Available in multiple pack sizes, including 50 mg.
  • Commonly used for studies of M4 signaling and antipsychotic-like activity in preclinical models.
Catalog No. 50-002-34776 Supplier Medchemexpress LLC Supplier No. HY12018450MG
VU0467485 (AZ13713945) is a potent, selective, orally bioavailable positive allosteric modulator of the muscarinic acetylcholine receptor M4, provided as a research reagent for receptor pharmacology and preclinical studies.
  • Potent and selective M4 positive allosteric modulator.
  • Orally bioavailable properties support in vivo evaluation.
  • High purity suitable for pharmacological assays.
  • Available in multiple pack sizes, including 50 mg.
  • Commonly used for studies of M4 signaling and antipsychotic-like activity in preclinical models.