Learn More
Medchemexpress LLC 5-amino-N-[(3-fluoro-4-methoxyphenyl)methyl]-3,4-dimethylthieno[2,3-c]pyridazine-6-carboxamide | 1451994-10-7 | 99.3% | 100 MG
SDP
Catalog No. 5000234551
Encompass_Preferred
VU0467485 (AZ13713945) is a potent, selective, orally bioavailable positive allosteric modulator of the muscarinic acetylcholine receptor M4. It is supplied for research use and exhibits CNS penetration and antipsychotic-like activity in preclinical models.
  • Potent M4 positive allosteric modulator
  • Selective over M1, M2, M3, and M5 receptors
  • Orally bioavailable with CNS penetration
  • High purity suitable for research applications
  • Provided as a 100 mg pack for assay and preclinical studies
Catalog No. 50-002-34551 Supplier Medchemexpress LLC Supplier No. HY120184100MG
VU0467485 (AZ13713945) is a potent, selective, orally bioavailable positive allosteric modulator of the muscarinic acetylcholine receptor M4. It is supplied for research use and exhibits CNS penetration and antipsychotic-like activity in preclinical models.
  • Potent M4 positive allosteric modulator
  • Selective over M1, M2, M3, and M5 receptors
  • Orally bioavailable with CNS penetration
  • High purity suitable for research applications
  • Provided as a 100 mg pack for assay and preclinical studies