Learn More
Medchemexpress LLC 5-amino-N-[(3-fluoro-4-methoxyphenyl)methyl]-3,4-dimethylthieno[2,3-c]pyridazine-6-carboxamide | 1451994-10-7 | 99.3% | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1201845MG
Encompass_Preferred
VU0467485 is a potent, selective, and orally bioavailable positive allosteric modulator of the muscarinic acetylcholine receptor M4, supplied as a solid research compound for in vitro and in vivo pharmacology studies.
  • Potent M4 positive allosteric modulator.
  • High purity (99.29%).
  • Molecular formula C17H17FN4O2S and molecular weight 360.41 g·mol⁻1.
  • Light yellow to yellow solid appearance.
  • Available in small solid quantities for research, including 5 MG packaging.
Catalog No. 50-002-34498