Learn More
Biotium 5-Aminoallyl-dUTP, Sodium Salt, Lyophilized
Catalog No. 501964630
Encompass_Preferred
5-Aminoallyl-dUTP can be enzymatically incorporated into DNA. The resulting amine-containing DNA can be subsequently labeled with a fluorescent dye, biotin or other haptens via conventional peptide coupling method. This two-step method for labeling nucleic acids is considerably more economical than the one-step method using a prelabeled dUTP. The lyophilized powder form of 5-Aminoallyl-dUTP is suitable for long-term storage. Properties: White solid soluble in water; Store at -20⁰C; C12H17N3Na3O14P3; MW: 589.17.
Catalog No. 50-196-4630 Supplier Biotium Supplier No. 400201
5-Aminoallyl-dUTP can be enzymatically incorporated into DNA. The resulting amine-containing DNA can be subsequently labeled with a fluorescent dye, biotin or other haptens via conventional peptide coupling method. This two-step method for labeling nucleic acids is considerably more economical than the one-step method using a prelabeled dUTP. The lyophilized powder form of 5-Aminoallyl-dUTP is suitable for long-term storage. Properties: White solid soluble in water; Store at -20⁰C; C12H17N3Na3O14P3; MW: 589.17.