Learn More
AdipoGen 5-Aminoeosin
SDP
Supplier:  AdipoGen CDXA0055M010
Encompass_Preferred
Chemical. CAS 75900-75-3. Formula C20H9Br4NO5. MW 662.9. Synthetic. Fluorescent marker used extensively in microscopy studies. Can be used in photoluminometric immunoassays for the formation of conjugates with immunoreactants by fluorescent energy transfer.
Catalog No. 50-256-1849