Learn More
Medchemexpress LLC HY-D0022 1g Medchemexpress, 5-Aminofluorescein CAS:3326-34-9 Purity:>98%
SDP
Catalog No. 501933132
Encompass_Preferred
Medchemexpress, HY-D0022 1g 5-Aminofluorescein CAS:3326-34-9 5-Aminofluorescein (5-AF) is a fluorescence marker, covalently bound to human serum albumin. The excitation wavelength is 490 nm and the emission wavelength is 515 nm. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3132 Supplier Medchemexpress LLC Supplier No. HYD00221G
Medchemexpress, HY-D0022 1g 5-Aminofluorescein CAS:3326-34-9 5-Aminofluorescein (5-AF) is a fluorescence marker, covalently bound to human serum albumin. The excitation wavelength is 490 nm and the emission wavelength is 515 nm. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.