Learn More
AdipoGen 5-Aminofluorescein
SDP
Supplier:  AdipoGen CDXA0018M250
Encompass_Preferred
Chemical. CAS 3326-34-9. Formula C20H13NO5. MW 347.32. Synthetic. The uronic acid residues of all known glycosaminoglycuronans react wit 5-aminofluorescein to yield fluorescent derivatives, which show fluorescence characteristics identical to those of 5-acetamidofluorescein CDX-A0089. 5-aminofluorescein is also used to prepare FITC Isomer I CDX-F0011, CDX-F0020.
Catalog No. 50-256-1868