Learn More
AdipoGen 5-Aminofluorescein diacetate
SDP
Catalog No. 502561748
Encompass_Preferred
Chemical. CAS 195977-46-9. Formula C24H17NO7. MW 431.39. 5-Aminofluorescein diacetate is a non-fluorescent probe that after entering the cells can be cleaved by esterases to form 5-Aminofluorescein 5-AF a fluorescence marker with excitation wavelength at 490 nm and the emission wavelength at 515 nm. The compound can be used as a negative control for the fluorescent probe DAF-2 DA. 5-Aminofluorescein diacetate could also be used as a starting material or intermediate in synthesis of fluorescent probes.
Catalog No. 50-256-1748 Supplier AdipoGen Supplier No. CDXA0318G001
Chemical. CAS 195977-46-9. Formula C24H17NO7. MW 431.39. 5-Aminofluorescein diacetate is a non-fluorescent probe that after entering the cells can be cleaved by esterases to form 5-Aminofluorescein 5-AF a fluorescence marker with excitation wavelength at 490 nm and the emission wavelength at 515 nm. The compound can be used as a negative control for the fluorescent probe DAF-2 DA. 5-Aminofluorescein diacetate could also be used as a starting material or intermediate in synthesis of fluorescent probes.