Learn More
Abcam 5-Aminofluorescein (Fluoresceinamine, isomer I), Fluorescence marker, 1G
Catalog No. 502705305
Encompass_Preferred
MW 347.3 Da, Purity >95%. Fluorescence marker. Selectively targets malignant glioma tissue in vivo when covalently linked to human serum albumin at a molar ratio of 1:1without bleaching nor penetrating into surrounding brain edema or necrotic tissue. Blood-brain barrier permeable.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5305 Supplier Abcam Supplier No. AB1453051G
MW 347.3 Da, Purity >95%. Fluorescence marker. Selectively targets malignant glioma tissue in vivo when covalently linked to human serum albumin at a molar ratio of 1:1without bleaching nor penetrating into surrounding brain edema or necrotic tissue. Blood-brain barrier permeable.
The product is subject to the following: Abcam Restricted Use Statement