Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Aminoimidazole-4-carboxamide 1-beta-D-ribofuranoside, 98%, Thermo Scientific Chemicals

Catalog No. AAJ64720MF
Change view
Click to view available options
Quantity:
50 mg
200 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64720MF 50 mg
AAJ64720EX3 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ64720MF Supplier Thermo Scientific Chemicals Supplier No. J64720MF
Add to Cart

Activator of AMP-activated protein kinase (AMPK)

5-Aminoimidazole-4-carboxamide 1-beta-D-ribofuranoside is used as a cell permeable activator of AMP-activated protein kinase (AMPK), a metabolic master regulator that is activated in times of reduced energy availability (high cellular AMP:ATP ratios) and serves to inhibit anabolic processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Aminoimidazole-4-carboxamide 1-beta-D-ribofuranoside is used as a cell permeable activator of AMP-activated protein kinase (AMPK), a metabolic master regulator that is activated in times of reduced energy availability (high cellular AMP:ATP ratios) and serves to inhibit anabolic processes.

Solubility
Soluble in DMSO at 2mg/ml. Soluble in water or ethanol at less than 1mg/ml

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2627-69-2
Molecular Formula C9H14N4O5
Molecular Weight (g/mol) 258.23
MDL Number MFCD00869751
InChI Key RTRQQBHATOEIAF-UUOKFMHZSA-N
Synonym acadesine, aica-riboside, aicar, arasine, aica riboside, aic-riboside, acadesina, acadesinum, 5-amino-1-beta-d-ribofuranosyl-1h-imidazole-4-carboxamide, protara
PubChem CID 17513
ChEBI CHEBI:28498
IUPAC Name 5-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]imidazole-4-carboxamide
SMILES NC(=O)C1=C(N)N(C=N1)[C@@H]1O[C@H](CO)[C@@H](O)[C@H]1O

Specifications

Melting Point 214°C to 215°C
Color Brown
Quantity 50 mg
Merck Index 14,16
Solubility Information Soluble in DMSO at 2mg/ml; Soluble in water or ethanol at less than 1mg/ml
Formula Weight 258.23
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-Aminoimidazole-4-carboxamide 1-beta-D-ribofuranoside
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 220-097-5
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.