Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Aminoimidazole-4-carboxamide-1-D-ribofuranosyl 5-monophosphate, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013863 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3863 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3863 Supplier MilliporeSigma Supelco Supplier No. 0659525MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 3031-94-5
Linear Formula C9H15N4O8 P
Synonym AICAR monophosphate; N1-(beta-D-5′-Phosphoribofuranosyl)-5-aminoimidazole-4-carboxamide; NSC 283955; NSC 292227; ZMP
Type Analytical Standard
Quantity 25 mg
Formula Weight 338.21
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.