Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Aminoindan-1-one, 97%, Thermo Scientific™

Catalog No. NRB01003DE
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NRB01003DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NRB01003DE Supplier Thermo Scientific Chemicals Supplier No. NRB01003DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3470-54-0
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
InChI Key HODOSJNSRPXYBH-UHFFFAOYSA-N
Synonym 5-aminoindan-1-one, 5-amino-1-indanone, 5-amino-2,3-dihydro-1h-inden-1-one, 1h-inden-1-one, 5-amino-2,3-dihydro, 5-aminoindanone, 5-amino-1-oxoindane, 5-amino-indan-1-one, pubchem14476, 5-amino-1-indanone #, 5-azanyl-2,3-dihydroinden-1-one
PubChem CID 312890
IUPAC Name 5-amino-2,3-dihydroinden-1-one
SMILES C1CC(=O)C2=C1C=C(C=C2)N

Specifications

Assay Percent Range 97%
Quantity 5 g
Formula Weight 147.18
Percent Purity 97%
Chemical Name or Material 5-Aminoindan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.