Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Aminoisophthalic acid | 99-31-0 | MFCD00002522 | 99.0% | 181.15 g·mol⁻¹ | C8H7NO4 | 100 G
SDP

Catalog No. 5000301115
Encompass_Preferred

5-Aminoisophthalic acid is an aromatic organic reagent used as a building block and biochemical reagent for life-science research, including in vitro proteomics applications. It is supplied as a solid suitable for synthesis and analytical workflows.

  • CAS number: 99-31-0.
  • Molecular formula: C8H7NO4.
  • Molecular weight: 181.15 g·mol⁻¹.
  • Purity: 99.0% (manufacturer listed).
  • Appearance: off-white to light yellow solid.
  • Available pack sizes: 25 g, 100 g, >100 g.
  • Storage: room temperature (3 years); in solution -80°C (2 years), -20°C (1 year).
  • Intended use: research use only; not for clinical use.

Catalog No. 50-003-01115 Supplier Medchemexpress LLC Supplier No. HYW016454100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Aminoisophthalic acid is an aromatic organic reagent used as a building block and biochemical reagent for life-science research, including in vitro proteomics applications. It is supplied as a solid suitable for synthesis and analytical workflows.

  • CAS number: 99-31-0.
  • Molecular formula: C8H7NO4.
  • Molecular weight: 181.15 g·mol⁻¹.
  • Purity: 99.0% (manufacturer listed).
  • Appearance: off-white to light yellow solid.
  • Available pack sizes: 25 g, 100 g, >100 g.
  • Storage: room temperature (3 years); in solution -80°C (2 years), -20°C (1 year).
  • Intended use: research use only; not for clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.