Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-aminoisophthalic acid | 99-31-0 | MFCD00002522 | 99.9% | 181.15 g/mol | C8H7NO4 | 25 G
SDP

Catalog No. 5000302760
Encompass_Preferred

5-Aminoisophthalic acid is an organic biochemical reagent (5-aminobenzene-1,3-dicarboxylic acid) used as a building block and reagent in organic synthesis and biochemical research. It is supplied as an off-white to light-yellow solid with high purity and good solubility in DMSO, and documentation is provided for safe handling and quality assurance.

  • High purity suitable for research applications.
  • Molecular weight 181.15 g/mol and formula C8H7NO4 for accurate stoichiometry.
  • Off-white to light-yellow solid that is easy to handle.
  • Soluble in DMSO (≥200 mg/mL) for convenient stock solution preparation.
  • Certificate of analysis, SDS, and handling instructions available for quality and safety.
  • Available in lab-scale quantities such as 25 g and 100 g for small-batch synthesis.

Catalog No. 50-003-02760 Supplier Medchemexpress LLC Supplier No. 25GHYW016454
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Aminoisophthalic acid is an organic biochemical reagent (5-aminobenzene-1,3-dicarboxylic acid) used as a building block and reagent in organic synthesis and biochemical research. It is supplied as an off-white to light-yellow solid with high purity and good solubility in DMSO, and documentation is provided for safe handling and quality assurance.

  • High purity suitable for research applications.
  • Molecular weight 181.15 g/mol and formula C8H7NO4 for accurate stoichiometry.
  • Off-white to light-yellow solid that is easy to handle.
  • Soluble in DMSO (≥200 mg/mL) for convenient stock solution preparation.
  • Certificate of analysis, SDS, and handling instructions available for quality and safety.
  • Available in lab-scale quantities such as 25 g and 100 g for small-batch synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.