Learn More
Selleck Chemical LLC 5-Aminolevulinic acid HCl 50mg 5451-09-2
SDP
Supplier:  Selleck Chemical LLC S255350MG
Encompass_Preferred
5-Aminolevulinic Acid HCl is an intermediate in the porphyrin synthesis pathway, used as a photosensitizing agent and a antineoplastic agent.
Catalog No. 50-508-5