Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(Aminomethyl)-4-methylthiazole, 97%, Thermo Scientific™

Catalog No. AC429650050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC429650050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC429650050 Supplier Thermo Scientific Chemicals Supplier No. 429650050

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 857549-15-6
Molecular Formula C5H8N2S
Molecular Weight (g/mol) 128.193
MDL Number MFCD08060724
InChI Key UGNFDYLWMWQMPR-UHFFFAOYSA-N
Synonym 4-methyl-1,3-thiazol-5-yl methanamine, 4-methylthiazol-5-yl methanamine, 4-methyl-5-thiazolemethanamine, 5-thiazolemethanamine, 4-methyl, 5-aminomethyl-4-methylthiazole, 1-4-methyl-1,3-thiazol-5-yl methanamine, 4-methyl-1,3-thiazol-5-yl methyl amine dihydrochloride, 1-4-methyl-1,3-thiazol-5-yl methanamine dihydrochloride, c-4-methyl-thiazol-5-yl-methylamine, 4-methyl-1,3-thiazol-5-yl methylamine
PubChem CID 24688991
IUPAC Name (4-methyl-1,3-thiazol-5-yl)methanamine
SMILES CC1=C(SC=N1)CN

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 5 g
Formula Weight 128.2
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 5-(Aminomethyl)-4-methylthiazole
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.