Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Aminooxindole 98.0+%, TCI America™
SDP

Catalog No. A26681G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26681G 1 g
A26685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A26681G Supplier TCI America Supplier No. A26681G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20876-36-2
Molecular Formula C8H8N2O
Molecular Weight (g/mol) 148.165
MDL Number MFCD02179603
InChI Key JPUYXUBUJJDJNL-UHFFFAOYSA-N
Synonym 5-Amino-2-indolinone
PubChem CID 2773213
IUPAC Name 5-amino-1,3-dihydroindol-2-one
SMILES C1C2=C(C=CC(=C2)N)NC1=O

Specifications

Melting Point 197°C
Quantity 1 g
Formula Weight 148.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Aminooxindole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.