Learn More
Medchemexpress LLC HY-15027 500mg Medchemexpress, 5-Aminosalicylic Acid CAS:89-57-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15027500MG
Encompass_Preferred
Medchemexpress, HY-15027 500mg 5-Aminosalicylic Acid CAS:89-57-6 5-Aminosalicylic acid (Mesalamine) acts as a specific PPARγ agonist and also inhibits p21-activated kinase 1 (PAK1) and NF-κB. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1672